Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 0.3330N ±0.0005N (0.333M), LabChem™

$71.71 - $168.80

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 98.69
CAS 1310-73-2
CAS Max % 1.31
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
UN Number UN1824
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC243104

 
lab chem
LC243104
Each for $71.71

LC243105

 
lab chem
LC243105
Each for $168.80
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 40
Storage Information Room Temperature
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Physical Form Liquid
Color Clear, colorless
Solubility Soluble in water
Linear Formula NaOH
Density 1.02g/mL
Concentration 0.3330N ±0.0005N (0.333M)
Chemical Name or Material Sodium Hydroxide
Identification Passes Test
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
UN Number UN1824
CAS Max % 98.69
CAS 7732-18-5
Specific Gravity 1.02g/mL
CAS Max % 1.31
Packaging Poly Bottle
Health Hazard 1 Danger
CAS 1310-73-2
Traceability to NIST Traceable to NIST
Synonym Caustic Soda
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.