Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 0.0227N ±0.0002N (0.0227M), LabChem™

$42.51 - $273.46

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 99.91
CAS 1310-73-2
CAS Max % 0.09
Purity Grade Certified Grade
Linear Formula NaOH
Formula Weight 40
Synonym Caustic Soda
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC242301

 
lab chem
LC242301
500mL Each for $42.51

LC242302

 
lab chem
LC242302
1L Each for $49.51

LC242305

 
lab chem
LC242305
20L Each for $273.46
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 40
Quantity 500mL
Decomposition Information Sodium oxide
Viscosity 1.01cSt
Chemical Name or Material Sodium Hydroxide
Concentration 0.0227N ±0.0002N (0.0227M)
Physical Form Liquid
Density 1g/mL
Linear Formula NaOH
CAS 7732-18-5
Identification Passes Test
Synonym Caustic Soda
Packaging Poly Bottle
CAS Max % 99.91
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 1310-73-2
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Solubility Soluble in water
Storage Information Room Temperature
Specific Gravity 1g/mL
CAS Max % 0.09
Traceability to NIST Traceable to NIST
Color Clear, colorless
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.