missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide, beads, 99.99% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4578006

 View more versions of this product

Catalog No. AA4578006



Specifications

Specifications

Sodium hydroxide
99.99% (metals basis)
MFCD00003548
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
2.13g/cm3 at 20°C
3mm Hg at 800°C
5g
1310-73-2
HNaO
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Solid
Odorless
White

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 215-185-5

Documents