Please login to your online account to display your discounted pricing

Sodium Hydroxide ACS AR Pellet, Macron Fine Chemicals™


macron

Quantity 12kg
Packaging Poly Pail

Manufacturer: avantor performance  770888

 View more versions of this product

Catalog No. MK7708P12

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Packaging Poly Pail
Quantity 12kg
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Purity Grade ACS
Physical Form Pellets
Molecular Formula NaOH
Formula Weight 40g/mol

Specifications

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Purity Grade ACS
Physical Form Pellets
Molecular Formula NaOH
Formula Weight 40g/mol