missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydroxide, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC451130050
Specifications
Sodium Hydroxide | |
HNaO | |
aetznatron, ascarite, caustic soda, soda lye, soda, caustic, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Extra Pure | |
Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone | |
1390.0°C |
1310-73-2 | |
NaOH | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
Plastic Bottle | |
318.0°C | |
5kg |
Chemical Identifiers
1310-73-2 | |
39.997 | |
aetznatron, ascarite, caustic soda, soda lye, soda, caustic, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |