missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hydroxide, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Manufacturer:  Alfa Aesar™ 88623LS

 View more versions of this product

Catalog No. AA88623LS



Specifications

Specifications

Sodium hydroxide Acculute Standard Volumetric Solution
1310-73-2
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Liquid
6 units
Final Concentration 0.1N
HNaO
UN1824
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Odorless

Chemical Identifiers

1310-73-2
39.997
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
HNaO
MFCD00003548
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION

TSCA : TSCA

SDS Documents