missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, 99 to 100%, ACS, EMD Millipore

$127.37 - $127.37

Chemical Identifiers

CAS 1310-73-2
Physical Form Pellet
Assay Percent Range 99 to 100%
Molecular Formula NaOH
Formula Weight 40
Synonym Caustic Soda
Chemical Name or Material Sodium Hydroxide, For ACS analysis
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MSX05901 Emd Millipore Corporation
SX05901
500g Each for $127.37
Specifications Description & Specifications

Chemical Identifiers

1310-73-2
99 to 100%
40
Sodium Hydroxide, For ACS analysis
Pellet
NaOH
Caustic Soda

Specifications

1310-73-2
99 to 100%
40
Plastic Bottle
Sodium Hydroxide, For ACS analysis
Pellet
NaOH
Caustic Soda
500g