Learn More
Sodium hydroxide, 98%, extra pure, pellets, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 134070100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hydroxide | |
1390.0°C | |
318.0°C | |
HNaO | |
MFCD00003548 | |
2.13g/mL | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
10 kg | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
98% | |
Plastic drum |
Beige to Brown | |
1310-73-2 | |
98% | |
NaOH | |
01,1083; 05,616; 07,336; 08,460 | |
15, 8761 | |
Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
Extra Pure | |
Crystalline Powder |
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
HNaO | |
MFCD00003548 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Keep only in original container. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only