missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC450580025

 View more versions of this product

Catalog No. AC450580025



Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Sodium Hydroxide
98%
NaOH
aetznatron, ascarite, caustic soda, soda lye, soda, caustic, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic
2.5kg
[OH-].[Na+]
39.997
CHEBI:32145
98%
Plastic Bottle
1310-73-2
HNaO
1390.0°C
318.0°C
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
Pure
Solubility in water: completely soluble. Other solubilities: soluble in alcohol and glycerol, insoluble in acetone

Chemical Identifiers

1310-73-2
39.997
aetznatron, ascarite, caustic soda, soda lye, soda, caustic, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
SDS Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only