Please login to your online account to display your discounted pricing

Sodium Hydroxide, 97.0% min., ACS, EMD Millipore


Quantity 500g

Manufacturer: emd millipore corporation  SX05931

 View more versions of this product

Catalog No. MSX05931

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Pellet
Purity Grade ACS Grade
Formula Weight 40
Quantity 500g
Packaging Plastic Bottle
Synonym Caustic Soda
Linear Formula NaOH
CAS 1310-73-2
Assay Percent Range 97.0%, min.
Chemical Name or Material Sodium Hydroxide
Molecular Formula NaOH
Name Note For ACS analysis

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note For ACS analysis
CAS 1310-73-2
Purity Grade ACS Grade
Physical Form Pellet
Assay Percent Range 97.0%, min.
Molecular Formula NaOH
Linear Formula NaOH
Formula Weight 40
Synonym Caustic Soda