missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, 97.0% min., ACS, EMD Millipore

$110.31 - $233.83

Chemical Identifiers

CAS 1310-73-2
Physical Form Pellet
Assay Percent Range 97.0%, min.
Molecular Formula NaOH
Linear Formula NaOH
Formula Weight 40
Synonym Caustic Soda
Chemical Name or Material Sodium Hydroxide, For ACS analysis
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MSX05931 Emd Millipore Corporation
SX05931
500g Each for $110.31
MSX05933 Emd Millipore Corporation
SX05933
2.5kg Each for $233.83
Specifications Description & Specifications

Chemical Identifiers

1310-73-2
97.0%, min.
NaOH
Caustic Soda
ACS Grade
Pellet
NaOH
40
Sodium Hydroxide, For ACS analysis

Specifications

1310-73-2
97.0%, min.
NaOH
Caustic Soda
500g
ACS Grade
Pellet
NaOH
40
Plastic Bottle
Sodium Hydroxide, For ACS analysis