Please login to your online account to display your discounted pricing

Sodium Hydroxide, 6.64 Normal (21.5% w/w), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2 (Sodium Hydroxide)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R74676501A

 
ricca chemical company
RIR74676501A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R74676504A

 
ricca chemical company
RIR74676504A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2 (Sodium Hydroxide)
Concentration or Composition Notes 6.64Normal,21.5% (w/w)
CAS 7732-18-5 (Water, Deionized)
Packaging Poly natural