missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified Reagent Grade, 50.0% (w/w) ±1.0%, LabChem™

Manufacturer:  LabChem LC241504

 View more versions of this product

Catalog No. LC241504



Specifications

Specifications

Sodium Hydroxide
4L
50,50
HNaO
UN1824
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Liquid
Certified Reagent
Soluble in water
1525kg/m3
12°C
Colorless
1310-73-2,7732-18-5
1525kg/m3
NaOH
Passes Test
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
50.0% ± 1.0% (w/w)
Poly Bottle
Room Temperature
Traceable to NIST
143°C

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If swallowed: Rinse

Danger

EINECSNumber : 215-185-5

Documents
Promotions

Promotions