Please login to your online account to display your discounted pricing

Sodium hydroxide, 50% w/w aq. soln., Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sodium hydroxide
CAS 1310-73-2
Molecular Formula NaOH
Formula Weight 40
MDL Number MFCD00003548
UN Number UN1824
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA3338222

 
alfa aesar
3338222
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 40
Quantity 100g
Chemical Name or Material Sodium hydroxide
TSCA TSCA
Physical Form Liquid
Molecular Formula NaOH
MDL Number MFCD00003548
CAS 1310-73-2
UN Number UN1824
Color Colorless
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314;
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration 50% w/w aq. soln.
Density 1.53
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314;
Causes severe skin burns and eye damage.