missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydroxide, 50% w/w aq. soln., Alfa Aesar™

Chemical Identifiers

CAS 1310-73-2
Physical Form Liquid
Molecular Formula NaOH
Formula Weight 40
MDL Number MFCD00003548
Synonym Sodium hydroxide
Chemical Name or Material Sodium hydroxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA33382A1 SDS Alfa Aesar
33382A1
1kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA33382A7 SDS Alfa Aesar
33382A7
5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1310-73-2
NaOH
MFCD00003548
Sodium hydroxide
Liquid
40
Sodium hydroxide

Specifications

1310-73-2
NaOH
40
Sodium hydroxide
23 hPa (17mm Hg) at 20°C
1kg
50%
Liquid
Colorless liquid
MFCD00003548
1.53g/mL
Colorless
Sodium hydroxide

Safety and Handling

H314: Causes severe skin burns and eye damage.

DOTInformation : DOT Class: 8; Packing Group: II

EINECSNumber : 215-185-5