missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, Certified, 50% (w/v) ±1%, LabChem™
Supplier: LabChem LC241404
Specifications
Sodium Hydroxide | |
1310-73-2,7732-18-5 | |
HNaO | |
UN1824 | |
Soluble in water | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
Certified | |
Poly Bottle | |
Liquid |
Colorless | |
64.03,35.97 | |
NaOH | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
4 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
50% ±1% (w/v) | |
Passes Test | |
Traceable to NIST |
Chemical Identifiers
1310-73-2 | |
39.997 | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto
Danger
Recommended Storage : Room Temperature