missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Hydroxide, 50% (w/v), Analytical Reagent Grade, Ricca Chemical
Suitable for Orsat Gas Analysis
Supplier: Ricca Chemical Company 72835HP
Specifications
Sodium Hydroxide | |
49.0,48.61 | |
HNaO | |
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
Analytical Reagent | |
Aqueous Solution |
1310-73-2,7732-18-5 | |
51.0,50.6 | |
50% (w/v) | |
20 L | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Hard Pack |