missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, 50% (w/v) Aq Soln, Anal. Reag Gr, for Orsat Gas Analysis, Ricca Chemical

$121.88 - $411.06

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
Grade Analytical Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
7283-1 Ricca Chemical Company
RI72831
Plastic 4L Each for $147.85
7283-32 Ricca Chemical Company
RI728332
1L Each for $121.88
7283-5 Ricca Chemical Company
RI72835
Cubitainer 20L Each for $411.06
Specifications

Description

For Orsat Gas Analysis
Description & Specifications

Chemical Identifiers

Sodium Hydroxide
Analytical Reagent Grade

Specifications

4L
Plastic
Analytical Reagent Grade
50% (w/v) Aqueous Solution
Sodium Hydroxide