Please login to your online account to display your discounted pricing

Sodium Hydroxide, 5.39 Normal (18% w/w), Ricca Chemical

$286.91 - $286.91

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 68.0
CAS Max % 71.0
CAS 1310-73-2
CAS Min % 29.6
CAS Max % 30.81
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R74653904A

 
ricca chemical company
RIR74653904A
Poly natural 4L Each for $286.91

R74653901A

 
ricca chemical company
RIR74653901A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 4L
Concentration or Composition Notes 5.39Normal,18% (w/w)
CAS Max % 71.0
CAS 1310-73-2
Chemical Name or Material Sodium Hydroxide
CAS Min % 29.6
CAS Max % 30.81
CAS Min % 68.0
Packaging Poly natural
CAS 7732-18-5