Please login to your online account to display your discounted pricing

Sodium Hydroxide, 5.00 Normal, Ricca Chemical

$49.58 - $600.15

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 70.0
CAS Max % 73.0
CAS 1310-73-2
CAS Min % 28.08
CAS Max % 29.22
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7465-1

  Accessories
ricca chemical company
RI74651
Poly natural 4L Each for $171.73

7465-32

  Accessories
ricca chemical company
RI746532
Poly natural 1L Each for $67.97

74652-5

 
ricca chemical company
RI74652.5
Cubitainer™ 10L Each for $358.27

74655

 
ricca chemical company
RI74655
Cubitainer™ 20L Each for $600.15

746516

 
ricca chemical company
RI746516
Poly natural 500mL Each for $49.58
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 4L
Concentration or Composition Notes 5.00N
CAS Min % 70.0
CAS Max % 73.0
Packaging Poly natural
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS 1310-73-2
CAS Min % 28.08
CAS Max % 29.22