Please login to your online account to display your discounted pricing

Sodium Hydroxide, 4N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$102.14 - $195.47

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide, 4N Volumetric Solution
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004139

 
avantor performance
566902
Narrow Mouth Polyethylene Bottle 1L Each for $102.14

02004140

 
avantor performance
566903
Cubetainer 4L Each for $195.47
Description & Specifications

Specifications

Chemical Name or Material Sodium Hydroxide, 4N Volumetric Solution
Quantity 1L
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.15kg