Please login to your online account to display your discounted pricing

Sodium Hydroxide, 40% (w/w) Aqueous Solution, Analytical Reagent Grade, Ricca Chemical

$46.48 - $46.48

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
CAS Min % 52.0
CAS Max % 55.0
CAS 7732-18-5
CAS Min % 45.6
CAS Max % 47.46
Purity Grade Analytical Reagent Grade
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

728816

  SDS
ricca chemical company
RI728816
Poly natural 500mL Each for $46.48

72885HP

 
ricca chemical company
RI72885HP
Ropack 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Purity Grade Analytical Reagent Grade
Concentration or Composition Notes 40% (w/v)
Solvent or Matrix Aqueous Solution
CAS Max % 55.0
CAS Min % 52.0
Packaging Poly natural
CAS 7732-18-5
CAS Max % 47.46
CAS Min % 45.6
CAS 1310-73-2
Chemical Name or Material Sodium Hydroxide