Please login to your online account to display your discounted pricing

Sodium Hydroxide, 30% (w/v) Aqueous Solution, Ricca Chemical


Quantity 20L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR727800020F

 View more versions of this product

Catalog No. R727800020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
Name Note 30% (w/v)
CAS 7732-18-5
CAS Min % 75.0
CAS Max % 78.0
CAS 1310-73-2
CAS Min % 22.69
CAS Max % 23.61
Packaging Cubitainer
Concentration or Composition Notes 30% (w/v)
Solvent or Matrix Aqueous Solution
Quantity 20L

Specifications

Chemical Name or Material Sodium Hydroxide
Name Note 30% (w/v)
CAS 7732-18-5
CAS Min % 75.0
CAS Max % 78.0
CAS 1310-73-2
CAS Min % 22.69
CAS Max % 23.61