Please login to your online account to display your discounted pricing

Sodium Hydroxide, 25% (w/w) Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$115.44 - $286.08

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide, 25% (w/w) Solution
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004134

 
avantor performance
566102
Narrow Mouth Polyethylene Bottle 1L Each for $115.44

02004135

 
avantor performance
566103
Cubetainer 4L Each for $286.08
Description & Specifications

Specifications

Formula Weight 40
Quantity 1L
Chemical Name or Material Sodium Hydroxide, 25% (w/w) Solution
Molecular Formula NaOH
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Packaging Narrow Mouth Polyethylene Bottle
Specific Gravity 1L = 1.27kg