missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, Certified, 20.0% (w/v) ±0.5% (w/v), LabChem™

Manufacturer:  LabChem LC240904

 View more versions of this product

Catalog No. LC240904



Specifications

Specifications

Sodium Hydroxide
4L
83.12,16.88
NaOH
UN1824
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
[OH-].[Na+]
39.997
CHEBI:32145
Liquid
Certified
Soluble in water
1.18g/mL
Colorless
1310-73-2,7732-18-5
HNaO
1.18g/mL
Passes Test
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
20.0% ± 0.5% (w/v)
Poly Bottle
Room Temperature
Traceable to NIST

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto

Danger

Documents
Promotions

Promotions