missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, 20% (w/v) Aqueous Solution, Ricca Chemical

Manufacturer:  Ricca Chemical Company RI72705

 View more versions of this product

Catalog No. 13-622-616



Specifications

Specifications

Sodium Hydroxide
1310-73-2,7732-18-5
85.0,17.01
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
Cubitainer™
20L
82.0,16.34
HNaO
20% (w/v) Aqueous Solution (200g/L)
[OH-].[Na+]
39.997
CHEBI:32145

Chemical Identifiers

1310-73-2
39.997
sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
CHEBI:32145
[OH-].[Na+]
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
14798
sodium;hydroxide
Documents
Promotions

Promotions