Please login to your online account to display your discounted pricing

Sodium Hydroxide, 2.50 Normal, Ricca Chemical

$44.67 - $44.67

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 1310-73-2
CAS Min % 9.08
CAS Max % 9.45
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

745716

  SDS
ricca chemical company
RI745716
Poly natural 500mL Each for $44.67

745755

 
ricca chemical company
RI745755
Drum 55g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Concentration or Composition Notes 2.50Normal
CAS Min % 89.0
CAS Max % 93.0
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
CAS 7732-18-5
Packaging Poly natural
CAS Min % 9.08
CAS Max % 9.45