Please login to your online account to display your discounted pricing

Sodium Hydroxide, 1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$74.70 - $148.99

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Name Note 1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004129

 
avantor performance
563502
Narrow Mouth Poly Bottle 1L Each for $74.70

02004130

 
avantor performance
563503
Cubetainer 4L Each for $148.99
Description & Specifications

Specifications

Chemical Name or Material Sodium Hydroxide
Quantity 1L
Name Note 1N Volumetric Solution
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
Packaging Narrow Mouth Poly Bottle
Storage Information Protect from air to avoid absorption of carbon dioxide
Specific Gravity 1L = 1.05kg