Please login to your online account to display your discounted pricing

Sodium Hydroxide, 12.0 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 50.0
CAS Max % 52.0
CAS 1310-73-2
CAS Min % 48.09
CAS Max % 50.06
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R74701201A

 
ricca chemical company
RIR74701201A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R74701204A

 
ricca chemical company
RIR74701204A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 1L
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 50.0
CAS 1310-73-2
CAS Min % 48.09
Concentration or Composition Notes 12.0Normal
CAS Max % 52.0
Packaging Poly natural
CAS Max % 50.06