Please login to your online account to display your discounted pricing

Sodium Hydroxide, 10% (w/w) Aqueous Solution, Ricca Chemical

$245.06 - $245.06

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 88.0
CAS Max % 92.0
CAS 1310-73-2
CAS Min % 9.97
CAS Max % 10.37
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

72612-5

  SDS
ricca chemical company
RI72612.5
Cubitainer 10L Each for $245.06

72615

 
ricca chemical company
RI72615
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

726155

 
ricca chemical company
RI726155
Drum 55g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 10L
Concentration or Composition Notes 10% (w/w)
Solvent or Matrix Aqueous Solution
CAS Min % 88.0
CAS Max % 92.0
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
Packaging Cubitainer
CAS 1310-73-2
CAS Max % 10.37
CAS Min % 9.97