Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hydroxide, 10.0N Standardized Solution

Chemical Identifiers

Chemical Name or Material Sodium hydroxide
CAS 1310-73-2
Name Note Standardized Solution
Formula Weight 40
MDL Number MFCD00003548
UN Number UN1824
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35635K2

 
alfa aesar™
35635K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 40
Chemical Name or Material Sodium hydroxide
TSCA TSCA
Color Colorless
Quantity 1L
Name Note Standardized Solution
Physical Form Liquid
Concentration 10.0N
Density 1.327g/mL
MDL Number MFCD00003548
CAS 1310-73-2
UN Number UN1824
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Odorless
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: SODIUM HYDROXIDE SOLUTION
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.