Please login to your online account to display your discounted pricing

Sodium Hydroxide, 1.0N Solution, Biotech Reagent, J.T.Baker™

Produced under GMP guidelines, but for which there is no USP/NF compendia for the product

$1,276.40 - $1,466.93

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide, 1.0N Solution
CAS 1310-73-2
Purity Grade Biotech Reagent Grade
Molecular Formula NaOH
Formula Weight 40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004119

 
avantor performance
032807
Poly Bottle 19L Each for $1,466.93

02004120

 
avantor performance
032808
Poly Drum 19L Each for $1,276.40
Description & Specifications

Specifications

Chemical Name or Material Sodium Hydroxide, 1.0N Solution
Quantity 19L
CAS 1310-73-2
Purity Grade Biotech Reagent Grade
Molecular Formula NaOH
Formula Weight 40
Packaging Poly Bottle
Specific Gravity 1L = 1.05kg