missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 1310-73-2 |
---|---|
Molecular Formula | HNaO |
Molecular Weight (g/mol) | 39.997 |
InChI Key | HEMHJVSKTPXQMS-UHFFFAOYSA-M |
Synonym | aetznatron, ascarite, caustic soda, soda lye, soda, caustic, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic |
PubChem CID | 14798 |
ChEBI | CHEBI:32145 |
IUPAC Name | sodium;hydroxide |
SMILES | [OH-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
74-501-6CS
|
Ricca Chemical Company
RIR745000012X500 |
6L | Poly natural |
Each for $419.97
|
|
74-503-2CS
|
Ricca Chemical Company
RIR745000012X1A |
12L | Poly natural |
Each for $494.24
|
|
74-501-CS
|
Ricca Chemical Company
RIR74500004X4A |
16L | Poly natural |
Each for $374.79
|
|
74-505-5
|
Ricca Chemical Company
RI745055 |
55 gal. | Drum |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1310-73-2 | |
39.997 | |
aetznatron, ascarite, caustic soda, soda lye, soda, caustic, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
14798 | |
sodium;hydroxide |
Specifications
1310-73-2,7732-18-5 | |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
Liquid | |
Odorless | |
Aqueous Solution | |
6L |
1.000 ± 0.001 N at 20°C | |
aetznatron, ascarite, caustic soda, soda lye, soda, caustic, sodium hydrate, sodium hydroxide, sodium hydroxide na oh, sodium hydroxide solution, white caustic | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Poly natural | |
Solubility in Water: Infinite | |
Colorless | |
Sodium Hydroxide, 1.00 Normal |