Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.5N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$72.28 - $544.24

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide, 0.5N Volumetric Solution
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004117

 
avantor performance
563403
Cubetainer 4L Each for $544.24

02004115

 
avantor performance
563402
Narrow Mouth Polyethylene Bottle 1L Each for $72.28
Description & Specifications

Specifications

Formula Weight 40
Quantity 4L
Chemical Name or Material Sodium Hydroxide, 0.5N Volumetric Solution
CAS 1310-73-2
Molecular Formula NaOH
Packaging Cubetainer
Specific Gravity 1L = 1.02kg
Purity Grade BAKER ANALYZED™ Reagent