Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.500N, Ricca Chemical


Quantity 4L
Packaging Cubitainer

Manufacturer: ricca chemical company  RIR74000004F

 View more versions of this product

Catalog No. R74000004F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Clear colorless
Quantity 4L
Chemical Name or Material Sodium Hydroxide
CAS Max % 3.94
CAS Min % 94.0
Concentration or Composition Notes 0.500N
CAS Min % 3.79
CAS 1310-73-2
Packaging Cubitainer
CAS Max % 98.0
CAS 7732-18-5

Specifications

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 94.0
CAS Max % 98.0
CAS 1310-73-2
CAS Min % 3.79
CAS Max % 3.94
Physical Form Liquid