Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.500N (N/2) in Methanol, ASTM E 200 Reagent, Ricca Chemical

$115.76 - $115.76

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
Purity Grade ASTM E 200 Reagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7473-32

 
ricca chemical company
RI747332
Plastic 1L Each for $115.76

7473-1

 
ricca chemical company
RI74731
Plastic 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1L
Purity Grade ASTM E 200 Reagent
Chemical Name or Material Sodium Hydroxide
Concentration or Composition (by Analyte or Components) 0.500N (N/2) in Methanol
Packaging Plastic