Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.500 Normal (N/2) USP Grade, Ricca Chemical

$312.73 - $432.45

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
Name Note 0.500 Normal (N/2), Volumetric Solution
CAS 7732-18-5
CAS 1310-73-2
Purity Grade USP
Physical Form Liquid
Assay Percent Range 0.5000 ± 0.0002 N at 20°C
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

74-001-6CS

  SDS
ricca chemical company
RIR740000012X500
Poly natural 12 x 500mL Each for $352.21

74-001-CS

  SDS
ricca chemical company
RIR74000004X4A
Poly natural 4 x 4L Each for $312.73

74-003-2CS

  SDS
ricca chemical company
RIR740000012X1A
Poly natural 12 x 1L Each for $432.45
Description & Specifications

Specifications

Chemical Name or Material Sodium Hydroxide
Color Clear colorless
Quantity 12 x 500mL
Name Note 0.500 Normal (N/2), Volumetric Solution
CAS 7732-18-5
CAS 1310-73-2
Purity Grade USP
Physical Form Liquid
Assay Percent Range 0.5000 ± 0.0002 N at 20°C
Packaging Poly natural
Solvent or Matrix Aqueous Solution