Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.333 Normal, Ricca Chemical

$34.70 - $34.70

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 95.0
CAS Max % 99.0
CAS 1310-73-2
CAS Min % 2.55
CAS Max % 2.65
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

737716

  SDS
ricca chemical company
RI737716
Poly natural 500mL Each for $34.70

73772-5

  SDS
ricca chemical company
RI73772.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 500mL
Concentration or Composition Notes 0.333Normal
CAS Min % 95.0
CAS Max % 99.0
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS 1310-73-2
Packaging Poly natural
CAS Min % 2.55
CAS Max % 2.65