Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.3125 Normal, Suitable for USDA Acidity Testing in Citrus Products, Ricca Chemical

$246.36 - $246.36

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 1310-73-2
CAS Min % 1.23
CAS Max % 1.29
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

73742-5

 
ricca chemical company
RI73742.5
Cubitainer 10L Each for $246.36

737432

 
ricca chemical company
RI737432
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 10L
Concentration or Composition Notes 0.3125Normal
CAS Min % 97.0
CAS Max % 100.0
Chemical Name or Material Sodium Hydroxide
CAS Min % 1.23
CAS 7732-18-5
CAS 1310-73-2
Packaging Cubitainer
CAS Max % 1.29

Suitable For USDA Acidity Testing in Citrus Products