Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.2N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$80.44 - $532.85

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide, 0.2N Volumetric Solution
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004112

 
avantor performance
566502
Narrow Mouth Polyethylene Bottle 1L Each for $80.44

02004114

 
avantor performance
566503
Cubetainer 4L Each for $532.85
Description & Specifications

Specifications

Formula Weight 40
Quantity 1L
Chemical Name or Material Sodium Hydroxide, 0.2N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
CAS 1310-73-2
Packaging Narrow Mouth Polyethylene Bottle