Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.200 Normal, Ricca Chemical

$38.15 - $291.06

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 1310-73-2
CAS Min % 1.55
CAS Max % 1.61
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

736016

  SDS
ricca chemical company
RI736016
Poly natural 500mL Each for $38.15

7360-5

 
ricca chemical company
RI73605
Cubitainer 20L Each for $291.06

73602-5

 
ricca chemical company
RI73602.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Concentration or Composition Notes 0.200Normal
CAS Min % 96.0
CAS Max % 100.0
CAS 1310-73-2
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 1.61
CAS Min % 1.55
Packaging Poly natural