Please login to your online account to display your discounted pricing

Sodium hydroxide, 0.2 N standard solution, ACROS Organics™


Quantity 500mL
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  349685000

 View more versions of this product

Catalog No. AC349685000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 39.99
Quantity 500mL
Color Clear colorless
Merck Index 15, 8761
EINECS Number 215-185-5
TSCA TSCA
Concentration 0.1996 to 0.2004N
Density 1.0100g/mL
RTECS Number WB4900000
Fieser 01,1083; 05,616; 07,336; 08,460
Chemical Name or Material Sodium hydroxide
Molecular Formula HNaO
Linear Formula NaOH
MDL Number MFCD00003548
CAS 1310-73-2
Synonym Caustic soda
Health Hazard 1 Warning
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
CAS 7732-18-5
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Sodium hydroxide
CAS 1310-73-2
CAS 7732-18-5
Molecular Formula HNaO
Linear Formula NaOH
Formula Weight 39.99
MDL Number MFCD00003548
Synonym Caustic soda
Accessories

Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents