Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.1N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$77.36 - $497.63

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Name Note 0.1N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004109

 
avantor performance
563602
Narrow Mouth Poly Bottle 1L Each for $77.36

02004111

 
avantor performance
563603
Cubetainer 4L Each for $497.63
Description & Specifications

Specifications

Formula Weight 40
Quantity 1L
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2
Molecular Formula NaOH
Name Note 0.1N Volumetric Solution
Packaging Narrow Mouth Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 1.00kg