missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydroxide, 0.1N Standardized Solution, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 035625K7
Description
Sodium hydroxide, 0.1N standardized solution is used for volumetric analysis in laboratories.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hydroxide | |
4 L | |
MFCD00003548 | |
[OH-].[Na+] | |
39.997 | |
CHEBI:32145 | |
Sodium hydroxide: 0.4%; Water: 99.6% | |
Miscible with glycerol, ethanol and methanol; Immiscible with acetone and ether |
1310-73-2 | |
HNaO | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
sodium;hydroxide | |
14798 | |
40 | |
23 hPa (17mm Hg) at 20°C | |
Liquid |
Chemical Identifiers
1310-73-2 | |
39.997 | |
HEMHJVSKTPXQMS-UHFFFAOYSA-M | |
CHEBI:32145 | |
[OH-].[Na+] |
HNaO | |
MFCD00003548 | |
14798 | |
sodium;hydroxide |
Safety and Handling
EINECSNumber : 215-185-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only