Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.100N, Ricca Chemical

$40.06 - $248.88

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide Solution
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 1310-73-2
CAS Min % 0.78
CAS Max % 0.81
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7350-1

 
ricca chemical company
RI73501
Poly natural 4L Each for $92.39

7350-25

  SDS
ricca chemical company
RI73502.5
Cubitainer™ 10L Each for $210.03

7350-16

 
ricca chemical company
RI735016
Poly natural 500mL Each for $52.97

7350-32

  Accessories SDS
ricca chemical company
RI735032
Poly natural 1L Each for $40.06

7350-5

 
ricca chemical company
RI73505
Cubitainer™ 20L Each for $248.88

73501CT

 
ricca chemical company
RI73501CT
Cubitainer™ 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 0.100N (N/10)
CAS 1310-73-2
CAS Min % 0.78
CAS Max % 0.81
CAS Min % 97.0
CAS Max % 100.0
Packaging Poly natural
Chemical Name or Material Sodium Hydroxide Solution