missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.100 Normal (N/10), Ricca Chemical

$308.97 - $553.88

Chemical Identifiers

CAS 7732-18-5,1310-73-2
CAS Min % 97.0,0.78
CAS Max % 100.0,0.81
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
73-501-6CS SDS Ricca Chemical Company
RIR735000012X500
Poly natural 6L Each for $553.88
73-503-2CS SDS Ricca Chemical Company
RIR735000012X1A
Poly natural 12L Each for $437.36
73-501-CS SDS Ricca Chemical Company
RIR73500004X4A
Poly natural 16L Each for $308.97
RIC01002ST Ricca Chemical Company
RI73505HP
Ropack 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,1310-73-2
100.0,0.81
Sodium Hydroxide
97.0,0.78
Liquid

Specifications

Clear colorless
7732-18-5,1310-73-2
100.0,0.81
0.100 Normal (N/10)
Sodium Hydroxide
6L
97.0,0.78
Liquid
Poly natural

Safety and Handling

ShelfLife : 24 Months