Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.067 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 1310-73-2
CAS Min % 0.52
CAS Max % 0.54
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R73430001A

 
ricca chemical company
RIR73430001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R7343000500

 
ricca chemical company
RIR7343000500A
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 1L
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Max % 100.0
Concentration or Composition Notes 0.067Normal
CAS 1310-73-2
Packaging Poly natural
CAS Min % 97.0
CAS Max % 0.54
CAS Min % 0.52