missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hydroxide, 0.05N Standardized Solution

$62.53 - $130.15

Chemical Identifiers

CAS 1310-73-2
Formula Weight 40
MDL Number MFCD00003548
Chemical Name or Material Sodium hydroxide, Standardized Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35623K2 SDS Alfa Aesar™
35623K2
1L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA35623K7 SDS Alfa Aesar™
35623K7
4L Each for $62.53
AA35623M4 SDS Alfa Aesar™
35623M4
20L Each for $130.15
Specifications Description & Specifications

Chemical Identifiers

1310-73-2
40
MFCD00003548
Sodium hydroxide, Standardized Solution

Specifications

1310-73-2
0.05N
MFCD00003548
1L
Liquid
40
Odorless
Sodium hydroxide, Standardized Solution

Safety and Handling

TSCA : TSCA