Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.02N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 4L
Packaging Cubetainer

Manufacturer: avantor performance  565303

 View more versions of this product

Catalog No. 02004104

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 40
Quantity 4L
Chemical Name or Material Sodium Hydroxide, 0.02N Volumetric Solution
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
CAS 1310-73-2
Packaging Cubetainer
Specific Gravity 1L = 1.00kg

Specifications

Chemical Name or Material Sodium Hydroxide, 0.02N Volumetric Solution
CAS 1310-73-2
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula NaOH
Formula Weight 40