Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.0227 Normal, 1mL = 1mg CO, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 1310-73-2
CAS Min % 0.17
CAS Max % 0.17
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

73102-5

 
ricca chemical company
RI73102.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

731016

 
ricca chemical company
RI731016
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

73105

 
ricca chemical company
RI73105
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 10L
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
Concentration or Composition Notes 0.0227Normal
Equivalence 1 mL = 1 mg CO
CAS Min % 98.0
CAS Max % 100.0
CAS 1310-73-2
Packaging Cubitainer
CAS Min % 0.17
CAS Max % 0.17